(4-butoxy-3,5-dimethylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

294799-1G 294799-500MG 294799-5G
77468-354EA 668.47 USD
77468-354 77481-040 77491-412
(4-butoxy-3,5-dimethylphenyl)(methyl)sulfane ≥95%
(4-butoxy-3,5-dimethylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.3623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22373618
CAS Number: 1443345-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR