1-(4-butoxy-3,5-dimethylphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

294797-1G 294797-500MG 294797-5G
77468-350EA 415.09 USD
77468-350 77481-036 77491-408
1-(4-butoxy-3,5-dimethylphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(4-butoxy-3,5-dimethylphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₄H₁₇ClF₂O₂
MW: 290.7333 g/mol
Storage Temperature: Ambient
CAS Number: 3002532-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR