1-(4-butoxy-3,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

294795-1G 294795-500MG 294795-5G
77468-346EA 415.09 USD
77468-346 77481-032 77491-404
1-(4-butoxy-3,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-butoxy-3,5-dimethylphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₄H₁₇F₃O₂
MW: 274.2787 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889504
CAS Number: 2969435-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR