2-(4-Ethoxy-3,5-dimethylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

294790-1G 294790-500MG 294790-5G
77468-342EA 415.09 USD
77468-342 77481-028 77491-400
2-(4-Ethoxy-3,5-dimethylphenyl)propan-2-ol ≥95%
2-(4-Ethoxy-3,5-dimethylphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.2967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889502
CAS Number: 92101-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR