(4-Ethoxy-3,5-dimethylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

294786-1G 294786-500MG 294786-5G
77468-338EA 415.09 USD
77468-338 77481-024 77491-396
(4-Ethoxy-3,5-dimethylphenyl)(ethyl)sulfane ≥95%
(4-Ethoxy-3,5-dimethylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₈OS
MDL Number: MFCD34889498
CAS Number: 2969242-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR