(4-Ethoxy-3,5-dimethylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

294784-1G 294784-500MG 294784-5G
77468-334EA 415.09 USD
77468-334 77481-020 77491-392
(4-Ethoxy-3,5-dimethylphenyl)triethylsilane ≥95%
(4-Ethoxy-3,5-dimethylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₆H₂₈OSi
MW: 264.4784 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889496
CAS Number: 2969380-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR