(4-Ethoxy-3,5-dimethylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

294782-1G 294782-500MG 294782-5G
77468-330EA 668.47 USD
77468-330 77481-016 77491-388
(4-Ethoxy-3,5-dimethylphenyl)(methyl)sulfane ≥95%
(4-Ethoxy-3,5-dimethylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.3091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370324
CAS Number: 1443328-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR