2-Chloro-1-(4-ethoxy-3,5-dimethylphenyl)-2,2-difluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
294780-1G
294780-500MG
294780-5G
294780-25G
77468-326EA
415.09
USD
77468-326
77481-012
77491-384
77501-378
2-Chloro-1-(4-ethoxy-3,5-dimethylphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(4-ethoxy-3,5-dimethylphenyl)-2,2-difluoroethanone
Precaución: For research use only
Formula:
C₁₂H₁₃ClF₂O₂ MW: 262.6802 g/mol Storage Temperature: Ambient |
CAS Number:
3002515-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...