5-(Methylthio)-2,2'-bithiophene ≥95%

Supplier: AOBChem USA

Warning

294369-1G 294369-500MG 294369-5G 294369-25G
77468-322EA 415.09 USD
77468-322 77481-008 77491-380 77501-374
5-(Methylthio)-2,2'-bithiophene ≥95%
5-(Methylthio)-2,2'-bithiophene

Precaución: For research use only

Formula: C₉H₈S₃
MW: 212.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28100141
CAS Number: 99119-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR