2,2'-Bithiophene-5-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Carboxy-2,2'-bithiophene, [2,2'-Bithiophene]-5-carboxylic acid

294352-1G 294352-500MG 294352-5G 294352-25G
77468-318EA 88.58 USD
77468-318 77481-004 77491-376 77501-370
2,2'-Bithiophene-5-carboxylic acid ≥95%
2,2'-Bithiophene-5-carboxylic acid

Precaución: For research use only

Formula: C₉H₆O₂S₂
Storage Temperature: Ambient
MDL Number: MFCD00159571
CAS Number: 2060-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR