N,N-Dimethyl-4-(2-propen-1-yl)benzenamine ≥95%

Supplier: AOBChem USA

Warning

293691-1G 293691-500MG 293691-5G 293691-25G
77468-310EA 320.33 USD
77468-310 77480-996 77491-364 77501-360
N,N-Dimethyl-4-(2-propen-1-yl)benzenamine ≥95%
N,N-Dimethyl-4-(2-propen-1-yl)benzenamine

Precaución: For research use only

Formula: C₁₁H₁₅N
MW: 161.24 g/mol
Storage Temperature: Ambient
CAS Number: 51601-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR