[5-(2-Thienyl)-3-pyridyl]methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-(Thiophen-2-yl)pyridin-3-yl)methanol

29365-1G 29365-500MG 29365-5G 29365-25G
77468-306EA 256.47 USD
77468-306 77480-992 77491-360 77501-358
[5-(2-Thienyl)-3-pyridyl]methanol ≥97%
[5-(2-Thienyl)-3-pyridyl]methanol

Precaución: For research use only

Formula: C₁₀H₉NOS
MW: 191.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06410298
CAS Number: 393861-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR