4-butoxy-1-iodo-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

292743-1G 292743-500MG 292743-5G 292743-25G
77468-294EA 415.09 USD
77468-294 77480-980 77491-346 77501-346
4-butoxy-1-iodo-2-methylbenzene ≥95%
4-butoxy-1-iodo-2-methylbenzene

Precaución: For research use only

Formula: C₁₁H₁₅IO
Storage Temperature: Ambient
MDL Number: MFCD34888985
CAS Number: 1446097-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR