(4-Ethoxy-2-methylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

292735-1G 292735-500MG 292735-5G 292735-25G
77468-290EA 415.09 USD
77468-290 77480-976 77491-342 77501-342
(4-Ethoxy-2-methylphenyl)triethylsilane ≥95%
(4-Ethoxy-2-methylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₅H₂₆OSi
Storage Temperature: Ambient
MDL Number: MFCD34888978
CAS Number: 2969422-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR