3-Methyl-5-((3-methylbenzyl)oxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

292688-1G 292688-500MG 292688-5G 292688-25G
77468-286EA 415.09 USD
77468-286 77480-972 77491-338 77501-338
3-Methyl-5-((3-methylbenzyl)oxy)benzaldehyde ≥95%
3-Methyl-5-((3-methylbenzyl)oxy)benzaldehyde

Precaución: For research use only

Formula: C₁₆H₁₆O₂
MW: 240.2970 g/mol
MDL Number: MFCD34888948
CAS Number: 2969462-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR