2-Methyl-4-((4-methylbenzyl)oxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

292669-1G 292669-500MG 292669-5G 292669-25G
77468-282EA 415.09 USD
77468-282 77480-968 77491-334 77501-334
2-Methyl-4-((4-methylbenzyl)oxy)benzaldehyde ≥95%
2-Methyl-4-((4-methylbenzyl)oxy)benzaldehyde

Precaución: For research use only

Formula: C₁₆H₁₆O₂
MW: 240.2970 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34888930
CAS Number: 2969297-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR