1,2-Dihydroacenaphthylene-5-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

292616-1G 292616-5G 292616-25G
77468-278EA 60.77 USD
77468-278 77491-330 77501-330
1,2-Dihydroacenaphthylene-5-carboxylic acid ≥95%
1,2-Dihydroacenaphthylene-5-carboxylic acid

Precaución: For research use only

Formula: C₁₃H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00222369
CAS Number: 55720-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR