2-Methyl-4-(piperidin-1-yl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29261-1G 29261-500MG 29261-5G
77468-274EA 706.58 USD
77468-274 77480-964 77491-328
2-Methyl-4-(piperidin-1-yl)phenylboronic acid pinacol ester ≥97%
2-Methyl-4-(piperidin-1-yl)phenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₈H₂₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD24449445
CAS Number: 1366131-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR