Methyl 3,6-difluoro-2-Methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

29249-1G 29249-500MG 29249-5G
77468-270EA 572.68 USD
77468-270 77480-958 77491-322
Methyl 3,6-difluoro-2-Methylbenzoate ≥95%
Methyl 3,6-difluoro-2-Methylbenzoate

Precaución: For research use only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16250138
CAS Number: 1378775-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR