5-Bromo-2-(3-methoxyphenoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

29230-1G 29230-500MG 29230-5G 29230-25G
77468-266EA 97.85 USD
77468-266 77480-954 77491-318 77501-324
5-Bromo-2-(3-methoxyphenoxy)pyridine ≥95%
5-Bromo-2-(3-methoxyphenoxy)pyridine

Precaución: For research use only

Formula: C₁₂H₁₀BrNO₂
MW: 280.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14652048
CAS Number: 1249147-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR