3-Benzyloxy-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

29200-1G 29200-500MG 29200-5G
77468-262EA 580.92 USD
77468-262 77480-950 77491-314
3-Benzyloxy-2-methoxyphenylboronic acid ≥97%
3-Benzyloxy-2-methoxyphenylboronic acid

Precaución: For research use only

Formula: C₁₄H₁₅BO₄
Storage Temperature: Ambient
MDL Number: MFCD28806025
CAS Number: 2121513-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR