2-Chloro-1-(3,4-dichloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

291392-1G 291392-500MG 291392-5G 291392-25G
77468-258EA 415.09 USD
77468-258 77480-946 77491-310 77501-320
2-Chloro-1-(3,4-dichloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3,4-dichloro-2-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂Cl₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD34888118
CAS Number: 2969380-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR