4-(3,4-Dichloro-2-fluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

291388-1G 291388-500MG 291388-5G 291388-25G
77468-254EA 415.09 USD
77468-254 77480-942 77491-306 77501-316
4-(3,4-Dichloro-2-fluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3,4-Dichloro-2-fluorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₁H₁₁Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD34888116
CAS Number: 2969251-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR