1-(3,4-Dichloro-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

291386-1G 291386-500MG 291386-5G 291386-25G
77468-250EA 415.09 USD
77468-250 77480-938 77491-302 77501-312
1-(3,4-Dichloro-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(3,4-Dichloro-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₉H₂Cl₂F₆O
MW: 311.0079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34888114
CAS Number: 2969431-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR