(3,4-Dichloro-2-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

291384-1G 291384-500MG 291384-5G 291384-25G
77468-246EA 415.09 USD
77468-246 77480-934 77491-298 77501-308
(3,4-Dichloro-2-fluorophenyl)(ethyl)sulfane ≥95%
(3,4-Dichloro-2-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇Cl₂FS
Storage Temperature: Ambient
CAS Number: 3002537-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR