1-(3,4-Dichloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

291382-1G 291382-500MG 291382-5G 291382-25G
77468-242EA 415.09 USD
77468-242 77480-930 77491-294 77501-304
1-(3,4-Dichloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(3,4-Dichloro-2-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₃Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD34888111
CAS Number: 2969241-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR