(4-Chloro-2,3-difluorophenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

291376-1G 291376-500MG 291376-5G 291376-25G
77468-238EA 415.09 USD
77468-238 77480-926 77491-290 77501-300
(4-Chloro-2,3-difluorophenyl)triethylsilane ≥95%
(4-Chloro-2,3-difluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₂H₁₇ClF₂Si
MW: 262.7986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34888106
CAS Number: 2969473-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR