(4-Chloro-2,3-difluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

291370-1G 291370-500MG 291370-5G 291370-25G
77468-234EA 415.09 USD
77468-234 77480-922 77491-286 77501-296
(4-Chloro-2,3-difluorophenyl)(ethyl)sulfane ≥95%
(4-Chloro-2,3-difluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇ClF₂S
Storage Temperature: Ambient
CAS Number: 3002435-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR