Cyclopropyl(2,3-difluoro-4-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

291365-1G 291365-500MG 291365-5G 291365-25G
77468-230EA 415.09 USD
77468-230 77480-918 77491-280 77501-292
Cyclopropyl(2,3-difluoro-4-methoxyphenyl)methanol ≥95%
Cyclopropyl(2,3-difluoro-4-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₂F₂O₂
MW: 214.2085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34888098
CAS Number: 2969246-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR