5-Chloro-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29136-1G 29136-500MG 29136-5G 29136-25G
77468-222EA 341.96 USD
77468-222 77480-916 77491-282 77501-286
5-Chloro-2-fluoro-3-methylbenzaldehyde ≥97%
5-Chloro-2-fluoro-3-methylbenzaldehyde

Precaución: For research use only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD26687656
CAS Number: 1538090-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR