2-(4-(2,6-difluorophenoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: AOBChem USA

Warning

29045-1G 29045-500MG 29045-5G 29045-25G
77468-218EA 338.87 USD
77468-218 77480-906 77491-270 77501-280
2-(4-(2,6-difluorophenoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(4-(2,6-difluorophenoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For research use only

Formula: C₁₈H₁₉BF₂O₃
MW: 332.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384050
CAS Number: 1196396-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR