4-Bromo-6-(trifluoromethyl)-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-6-trifluoromethyl-1H-indazole

29024-1G 29024-5G 29024-25G
77468-214EA 60.77 USD
77468-214 77491-266 77501-276
4-Bromo-6-(trifluoromethyl)-1H-indazole ≥95%
4-Bromo-6-(trifluoromethyl)-1H-indazole

Precaución: For research use only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026956
CAS Number: 1000342-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR