5'-Iodo-m-terphenyl ≥95%

Supplier: AOBChem USA

Warning

290036-1G 290036-500MG 290036-5G 290036-25G
77468-210EA 95.79 USD
77468-210 77480-900 77491-262 77501-272
5'-Iodo-m-terphenyl ≥95%
5'-Iodo-m-terphenyl

Precaución: For research use only

Formula: C₁₈H₁₃I
Storage Temperature: Ambient
MDL Number: MFCD26131291
CAS Number: 87666-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR