[1,1':3',1''-terphenyl]-5'-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

290033-1G 290033-500MG 290033-5G 290033-25G
77468-206EA 415.09 USD
77468-206 77480-896 77491-258 77501-268
[1,1':3',1''-terphenyl]-5'-carbaldehyde ≥95%
[1,1':3',1''-terphenyl]-5'-carbaldehyde

Precaución: For research use only

Formula: C₁₉H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD30291184
CAS Number: 220955-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR