3-(Cyclopropylmethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

28773-1G 28773-500MG 28773-5G 28773-25G
77468-194EA 286.34 USD
77468-194 77480-886 77491-242 77501-248
3-(Cyclopropylmethoxy)benzaldehyde ≥97%
3-(Cyclopropylmethoxy)benzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06246720
CAS Number: 58986-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR