(4-Bromo-3-chloro-2-fluorophenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287172-1G 287172-500MG 287172-5G
77468-190EA 546.93 USD
77468-190 77480-882 77491-238
(4-Bromo-3-chloro-2-fluorophenyl)cyclopropylmethanone ≥95%
(4-Bromo-3-chloro-2-fluorophenyl)cyclopropylmethanone

Precaución: For research use only

Formula: C₁₀H₇BrClFO
MW: 277.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34885942
CAS Number: 1696258-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR