Cyclopropyl(2-fluoro-5-methoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287127-1G 287127-500MG 287127-5G
77468-186EA 546.93 USD
77468-186 77480-878 77491-234
Cyclopropyl(2-fluoro-5-methoxyphenyl)methanone ≥95%
Cyclopropyl(2-fluoro-5-methoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₁H₁₁FO₂
MW: 194.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26686474
CAS Number: 1519374-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR