Cyclopropyl(2-fluoro-5-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287117-1G 287117-500MG 287117-5G 287117-25G
77468-182EA 397.58 USD
77468-182 77480-874 77491-230 77501-242
Cyclopropyl(2-fluoro-5-methylphenyl)methanone ≥95%
Cyclopropyl(2-fluoro-5-methylphenyl)methanone

Precaución: For research use only

Formula: C₁₁H₁₁FO
MW: 178.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222563
CAS Number: 1342411-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR