Cyclopropyl(2-ethoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287100-1G 287100-500MG 287100-5G
77468-178EA 500.58 USD
77468-178 77480-870 77491-226
Cyclopropyl(2-ethoxyphenyl)methanone ≥95%
Cyclopropyl(2-ethoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17223680
CAS Number: 1339680-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR