Cyclopropyl(4-methoxy-3,5-dimethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287083-1G 287083-500MG 287083-5G
77468-174EA 540.75 USD
77468-174 77480-866 77491-222
Cyclopropyl(4-methoxy-3,5-dimethylphenyl)methanone ≥95%
Cyclopropyl(4-methoxy-3,5-dimethylphenyl)methanone

Precaución: For research use only

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24363272
CAS Number: 1528666-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR