Cyclopropyl[2-(1-methylethoxy)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

287063-1G 287063-500MG 287063-5G
77468-170EA 576.8 USD
77468-170 77480-862 77491-218
Cyclopropyl[2-(1-methylethoxy)phenyl]methanone ≥95%
Cyclopropyl[2-(1-methylethoxy)phenyl]methanone

Precaución: For research use only

Formula: C₁₃H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD23895259
CAS Number: 1526802-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR