Cyclopropyl(2-ethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287044-1G 287044-500MG 287044-5G 287044-25G
77468-166EA 382.13 USD
77468-166 77480-858 77491-214 77501-240
Cyclopropyl(2-ethylphenyl)methanone ≥95%
Cyclopropyl(2-ethylphenyl)methanone

Precaución: For research use only

Formula: C₁₂H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD28023817
CAS Number: 1600193-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR