(5-Bromo-2-fluorophenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287019-1G 287019-5G 287019-25G
77468-162EA 67.98 USD
77468-162 77491-210 77501-236
(5-Bromo-2-fluorophenyl)(cyclopropyl)methanone ≥95%
(5-Bromo-2-fluorophenyl)(cyclopropyl)methanone

Precaución: For research use only

Formula: C₁₀H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD18783211
CAS Number: 1222368-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR