5-(1,1-Dimethylethyl)-α-ethyl-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

286995-1G 286995-500MG 286995-5G
77468-158EA 676.71 USD
77468-158 77480-852 77491-206
5-(1,1-Dimethylethyl)-α-ethyl-2-methoxybenzenemethanol ≥95%
5-(1,1-Dimethylethyl)-α-ethyl-2-methoxybenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14530444
CAS Number: 35155-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR