4-[(1,1-Dimethylethyl)thio]-3-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

286529-1G 286529-500MG 286529-5G 286529-25G
77468-154EA 397.58 USD
77468-154 77480-848 77491-202 77501-232
4-[(1,1-Dimethylethyl)thio]-3-(trifluoromethyl)benzaldehyde ≥95%
4-[(1,1-Dimethylethyl)thio]-3-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₂H₁₃F₃OS
MW: 262.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23690643
CAS Number: 1554171-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR