2-Amino-5-chloro-3-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-3-methoxypyridin-2-amine

28532-1G 28532-500MG 28532-5G 28532-25G
77468-150EA 131.84 USD
77468-150 77480-844 77491-196 77501-226
2-Amino-5-chloro-3-methoxypyridine ≥97%
2-Amino-5-chloro-3-methoxypyridine

Precaución: For research use only

Formula: C₆H₇ClN₂O
MW: 158.586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015845
CAS Number: 1242336-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR