1-Ethoxy-4-methyl-2-(trimethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

284661-1G 284661-500MG 284661-5G 284661-25G
77468-146EA 415.09 USD
77468-146 77480-840 77491-192 77501-222
1-Ethoxy-4-methyl-2-(trimethylsilyl)benzene ≥95%
1-Ethoxy-4-methyl-2-(trimethylsilyl)benzene

Precaución: For research use only

Formula: C₁₂H₂₀OSi
MW: 208.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34885118
CAS Number: 1313182-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR