1-(5-Ethoxy-2-fluorophenyl)-1-phenylethanol ≥95%

Supplier: AOBChem USA

Warning

284349-1G 284349-500MG 284349-5G 284349-25G
77468-142EA 415.09 USD
77468-142 77480-836 77491-188 77501-218
1-(5-Ethoxy-2-fluorophenyl)-1-phenylethanol ≥95%
1-(5-Ethoxy-2-fluorophenyl)-1-phenylethanol

Precaución: For research use only

Formula: C₁₆H₁₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34885028
CAS Number: 2969485-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR