1-(6-Bromo-3-chloro-2-fluorophenyl)-3-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

284241-1G 284241-500MG 284241-5G 284241-25G
77468-138EA 415.09 USD
77468-138 77480-832 77491-184 77501-214
1-(6-Bromo-3-chloro-2-fluorophenyl)-3-methylbutan-1-ol ≥95%
1-(6-Bromo-3-chloro-2-fluorophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃BrClFO
MW: 295.5757 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34884962
CAS Number: 2969242-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR