4-Allyloxy-1-bromobenzene ≥95%

Supplier: AOBChem USA

Warning

28414-1G 28414-500MG 28414-5G 28414-25G
77468-134EA 112.27 USD
77468-134 77480-828 77491-180 77501-210
4-Allyloxy-1-bromobenzene ≥95%
4-Allyloxy-1-bromobenzene

Precaución: For research use only

Formula: C₉H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD00296908
CAS Number: 25244-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR